Learn More
Biomatik Corporation Fish Complement 3,C3 ELISA Kit
Supplier:  Biomatik Corporation EKC32147
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 12.5 μg/mL-500 μg/mL. Sensitivity: 12.5 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7262