Learn More
Biomatik Corporation Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit, 96 tests
Catalog No. 501503656
Encompass_Preferred
The Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit allows for quantitative determination of Prothrombin Fragment 1+2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 -50 ng/mL. Sensitivity: 0.35 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00734. Cross Activity: No significant cross-reactivity or interference between Prothrombin Fragment 1+2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3656 Supplier Biomatik Corporation Supplier No. EKU06964
The Human Prothrombin Fragment 1+2 (F1+2) ELISA Kit allows for quantitative determination of Prothrombin Fragment 1+2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 -50 ng/mL. Sensitivity: 0.35 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00734. Cross Activity: No significant cross-reactivity or interference between Prothrombin Fragment 1+2 and analogues was observed. Restriction: For Research Use Only.