Learn More
Biomatik Corporation Fish Interleukin 1Β (Il- 1Β) Elisa Kit
Supplier:  Biomatik Corporation EKC32160
Encompass_Preferred
Fish Interleukin 1Β (Il- 1Β) Elisa Kit for detecting Interleukin 1Β (Il- 1Β) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9YGD3; il1b. Target Synonyms: il1bInterleukin-1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7271