Learn More
Biomatik Corporation Fish Superoxide Dismutase (Sod) Elisa Kit
Supplier:  Biomatik Corporation EKC32167
Encompass_Preferred
Fish Superoxide Dismutase (Sod) Elisa Kit for detecting Superoxide Dismutase(Sod)Elisa Kit in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 1.25-20ng/ml. Sensitivity: 0.625ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: O73872; sod1. Target Synonyms: sod1; cuzn; Superoxide dismutase [Cu-Zn]; EC 1.15.1.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7276