Learn More
Biomatik Corporation Goat Haptoglobin,Hpt/HP ELISA KIT
Supplier:  Biomatik Corporation EKC32198
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Capra hircus (Goat). Detection Range: 3.9 ng/mL-1000 ng/mL. Sensitivity: 1.95 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7278