Learn More
Biomatik Corporation Goat Interferon Γ , Ifn-Γ Elisa Kit
Supplier:  Biomatik Corporation EKC32206
Encompass_Preferred
Goat Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon, Gamma in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P79154; IFNG. Target Synonyms: IFNG; Interferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7281