Learn More
Biomatik Corporation Human Pyruvate Kinase, Liver And RBC (PK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06992
Encompass_Preferred
The Human Pyruvate Kinase, Liver And RBC (PK) ELISA Kit allows for quantitative determination of Pyruvate Kinase, Liver And RBC in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 39-2,500 pg/mL. Sensitivity: 17 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P30613. Cross Activity: No significant cross-reactivity or interference between Pyruvate Kinase, Liver And RBC and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3662