Learn More
Biomatik Corporation Goat Interleukin 2 (Il-2) Elisa Kit
Supplier:  Biomatik Corporation EKC32211
Encompass_Preferred
Goat Interleukin 2 (Il-2) Elisa Kit for detecting Interleukin 2 in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36835; IL2. Target Synonyms: IL2Interleukin-2; IL-2; T-cell growth factor; TCGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7283