Learn More
Biomatik Corporation Mouse Pyruvate Kinase, Muscle (PKM2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06995
Encompass_Preferred
Mouse Pyruvate kinase isozymes M2 (PKM2) ELISA Kit for detecting PKM2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52480. Gene ID: 18746. Target Synonyms: M2PK; PKM; M2-PK; PKM1; CTHBP; OIP3; PK3; PKM; TCB; THBP1; Pyruvate kinase muscle isozyme; Thyroid hormone-binding protein 1; Cytosolic thyroid hormone-binding protein; Pyruvate Kinase, Muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3664