Learn More
Biomatik Corporation Guinea Pig Immunoglobulin E (Ige) Elisa Kit
Supplier:  Biomatik Corporation EKC32257
Encompass_Preferred
Guinea Pig Immunoglobulin E (Ige) Elisa Kit for detecting Immunoglobulin E (Ige) in serum, plasma, tissue homogenates. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 93.75-6000ng/ml. Sensitivity: 23.4ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7287