Learn More
Biomatik Corporation Human RAD51 Homolog (RAD51) ELISA Kit, 96 tests
Catalog No. 501503666
Encompass_Preferred
The Human RAD51 Homolog (RAD51) ELISA Kit allows for quantitative determination of RAD51 Homolog in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q06609. Cross Activity: No significant cross-reactivity or interference between RAD51 Homolog and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3666 Supplier Biomatik Corporation Supplier No. EKU07007
The Human RAD51 Homolog (RAD51) ELISA Kit allows for quantitative determination of RAD51 Homolog in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q06609. Cross Activity: No significant cross-reactivity or interference between RAD51 Homolog and analogues was observed. Restriction: For Research Use Only.