Learn More
Biomatik Corporation Rat Receptor Activator of Nuclear Factor Kappa B Ligand (RANkL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07021
Encompass_Preferred
Rat Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL) ELISA Kit for detecting RANkL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESE2. Gene ID: 117516. Target Synonyms: CD254; TNFSF11; ODF; OPGL; TRANCE; HRANKL2; SOdf; Tumor Necrosis Factor (ligand) superfamily Member 11; TNF-related activation-induced cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3669