Learn More
Biomatik Corporation Human Alpha1 Antichymotrypsin, Aact Elisa Kit
Supplier:  Biomatik Corporation EKC32535
Encompass_Preferred
Human Alpha1 Antichymotrypsin, Aact Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade A (Alpha-1 Antiproteinase, Antitrypsin), Member 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01011; SERPINA3. Target Synonyms: SERPINA3; AACT; AACT_HUMAN; ACT; alpha 1 Antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Antichymotrypsin; Cell growth inhibiting gene 24/25 protein; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; Growth inhibiting protein 24; Growth inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A (alpha 1 antiproteinase; antitrypsin) member 3; Serine (or cysteine) proteinase inhibitor clade A member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7309