Learn More
Biomatik Corporation Rat Receptor Interacting Serine Threonine Kinase 1 (RIPK1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07031
Encompass_Preferred
Rat Receptor Interacting Serine Threonine Kinase 1 (RIPK1) ELISA Kit for detecting RIPK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZYL0. Gene ID: 306886. Target Synonyms: RIP; Receptor-Interacting Protein; Cell death protein RIP; Serine/threonine-protein kinase RIP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3673