Learn More
Biomatik Corporation Human Receptor Tyrosine Kinase Like Orphan Receptor 1 (ROR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07034
Encompass_Preferred
Human Receptor Tyrosine Kinase Like Orphan Receptor 1 (ROR1) ELISA Kit for detecting ROR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01973. Gene ID: 4919. Target Synonyms: NTRKR1; Tyrosine-protein kinase transmembrane receptor ROR1; Neurotrophic tyrosine kinase, receptor-related 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3675