Learn More
Biomatik Corporation Human Reelin (RL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07035
Encompass_Preferred
Human Reelin (RELN) ELISA Kit for detecting RELN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P78509. Gene ID: 5649. Target Synonyms: RL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3676