Learn More
Biomatik Corporation Human Aryl Hydrocarbon Receptor, Ahr Elisa Kit
Supplier:  Biomatik Corporation EKC32794
Encompass_Preferred
Human Aryl Hydrocarbon Receptor, Ahr Elisa Kit for detecting Aryl Hydrocarbon Receptor in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35869; AHR. Target Synonyms: Ah receptor; AhR; AHR_HUMAN; Aromatic hydrocarbon receptor; Aryl hydrocarbon receptor; Aryl hydrocarbon receptor precursor; bHLHe76; Class E basic helix loop helix protein 76; Class E basic helix-loop-helix protein 76; HGNC:348. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7330