Learn More
Biomatik Corporation Human Regenerating Islet Derived Protein 3 Alpha (REG3a) ELISA Kit, 96 tests
Catalog No. 501503678
Encompass_Preferred
Human Regenerating Islet Derived Protein 3 Alpha (REG3a) ELISA Kit for detecting REG3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06141. Gene ID: 5068. Target Synonyms: HIP; INGAP; PAP-H; PAP1; PBCGF; REG-III; REG3; Pancreatitis-Associated Protein; Hepatointestinal pancreatic protein; Human proislet peptide; Pancreatitis-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3678 Supplier Biomatik Corporation Supplier No. EKU07043
Human Regenerating Islet Derived Protein 3 Alpha (REG3a) ELISA Kit for detecting REG3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06141. Gene ID: 5068. Target Synonyms: HIP; INGAP; PAP-H; PAP1; PBCGF; REG-III; REG3; Pancreatitis-Associated Protein; Hepatointestinal pancreatic protein; Human proislet peptide; Pancreatitis-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.