Learn More
Biomatik Corporation Human Basic Fibroblast Growth Factor, Bfgf Elisa Kit
Supplier:  Biomatik Corporation EKC32831
Encompass_Preferred
Human Basic Fibroblast Growth Factor, Bfgf Elisa Kit for detecting Basic Fibroblast Growth Factor, Bfgf in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09038; FGF2. Target Synonyms: Basic fibroblast growth factor; Basic fibroblast growth factor bFGF; BFGF; FGF 2; FGF B; FGF-2; Fgf2; FGF2 basic; FGF2_HUMAN; FGFB; Fibroblast growth factor 2 (basic); Fibroblast growth factor 2; Fibroblast growth factor; basic; HBGF 2; HBGF-2; HBGF2; HBGH 2; HBGH2; Heparin binding growth factor 2 precursor; Heparin-binding growth factor 2; Prostatropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7336