Learn More
Biomatik Corporation Mouse Regenerating Islet Derived Protein 3 Beta (REG3b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07046
Encompass_Preferred
Mouse Regenerating Islet Derived Protein 3 Beta (REG3b) ELISA Kit for detecting REG3b in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.4-600pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35230. Gene ID: 18489. Target Synonyms: PAP1; Pancreatitis-Associated Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3680