Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 8B (Bmp8B) Elisa Kit
Supplier:  Biomatik Corporation EKC32898
Encompass_Preferred
Human Bone Morphogenetic Protein 8B (Bmp8B) Elisa Kit for detecting Bone Morphogenetic Protein 8B in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 9.37-600pg/ml. Sensitivity: 2.34pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P34820; BMP8B. Target Synonyms: BMP-8; BMP-8B; BMP8; Bmp8b; BMP8B_HUMAN; bone morphogenetic protein 8 (osteogenic protein 2); Bone morphogenetic protein 8; Bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); OP-2; OP2; Osteogenic protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7345