Learn More
Biomatik Corporation Human Regenerating Islet Derived Protein 4 (REG4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07049
Encompass_Preferred
Human Regenerating Islet Derived Protein 4 (REG4) ELISA Kit for detecting REG4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BYZ8. Gene ID: 83998. Target Synonyms: GISP; REG-IV; RELP; Regenerating Gene Type IV; Gastrointestinal Secretory Protein; REG-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3683