Learn More
Biomatik Corporation Human Cartilage Oligomeric Protein, Comp Elisa Kit
Supplier:  Biomatik Corporation EKC33023
Encompass_Preferred
Human Cartilage Oligomeric Protein, Comp Elisa Kit for detecting Cartilage Oligomeric Matrix Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49747; COMP. Target Synonyms: cartilage oligomeric matrix protein (pseudoachondroplasia; epiphyseal dysplasia 1; multiple); Cartilage oligomeric matrix protein; Cartilage oligomeric matrix protein precursor; COMP; COMP_HUMAN; EDM 1; EDM1; EPD 1; EPD1; Epiphyseal dysplasia 1; Epiphyseal dysplasia 1 multiple; Epiphyseal dysplasia multiple 1; MED; MGC13181; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1; multiple); Pseudoachondroplasia; THBS 5; THBS5; Thrombospondin 5; Thrombospondin-5; Thrombospondin5; TSP5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7353