Learn More
Biomatik Corporation Human Catalase (Cat) Elisa Kit
Supplier:  Biomatik Corporation EKC33036
Encompass_Preferred
Human Catalase (Cat) Elisa Kit for detecting Catalase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04040; CAT. Target Synonyms: Cas1; CAT; CATA_HUMAN; Catalase; Cs1; MGC138422; MGC138424. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7355