Learn More
Biomatik Corporation Rat Relaxin (RLN) ELISA Kit, 96 tests
Catalog No. 501503686
Encompass_Preferred
Rat Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.79pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01347. Gene ID: 25616. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3686 Supplier Biomatik Corporation Supplier No. EKU07063
Rat Relaxin (RLN) ELISA Kit for detecting RLN in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.79pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01347. Gene ID: 25616. Target Synonyms: RLXH1; Relaxin H1; Prorelaxin H1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.