Learn More
Biomatik Corporation Human Chromogranin, Cga Elisa Kit
Supplier:  Biomatik Corporation EKC33132
Encompass_Preferred
Human Chromogranin, Cga Elisa Kit for detecting Chromogranin A (Parathyroid Secretory Protein 1) in serum, plasma, cell culture supernates, urine, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10645; CHGA. Target Synonyms: beta Granin; betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; CHG A; Chga; chromofungin; Chromogranin A; Chromogranin A parathyroid secretory protein 1; Chromogranin A precursor; ChromograninA; CMGA_HUMAN; ER-37; Pancreastatin; Parastatin; Parathyroid secretory protein 1; Pituitary secretory protein I; Secretory protein I; SP I; SP-I; SP1; SPI; vasostatin 2; Vasostatin; Vasostatin I; Vasostatin II; vasostatin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7360