Learn More
Biomatik Corporation Human Claudin-1 (Cldn1) Elisa Kit
Supplier:  Biomatik Corporation EKC33143
Encompass_Preferred
Human Claudin-1 (Cldn1) Elisa Kit for detecting Claudin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95832; CLDN1. Target Synonyms: CLDN1; CLD1; SEMP1; UNQ481/PRO944; Claudin-1; Senescence-associated epithelial membrane protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7363