Learn More
Biomatik Corporation Human Coagulation Factor Vii, Fvii Elisa Kit
Supplier:  Biomatik Corporation EKC33175
Encompass_Preferred
Human Coagulation Factor Vii, Fvii Elisa Kit for detecting Coagulation Factor Vii (Serum Prothrombin Conversion Accelerator) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08709; F7. Target Synonyms: coagulation factor VII (serum prothrombin conversion accelerator); Coagulation factor VII; Eptacog alfa; F7; FA7_HUMAN; Factor VII; Factor VII heavy chain; Factor VII light chain; FVII coagulation protein; OTTHUMP00000018733; OTTHUMP00000018734; Proconvertin; Serum prothrombin conversion accelerator; SPCA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7365