Learn More
Biomatik Corporation Human Collagen Alpha-1 (Ii) Chain (Col2A1) Elisa Kit
Supplier:  Biomatik Corporation EKC33194
Encompass_Preferred
Human Collagen Alpha-1 (Ii) Chain (Col2A1) Elisa Kit for detecting Collagen, Type Ii, Alpha 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02458; COL2A1. Target Synonyms: Alpha 1 type II collagen; Alpha-1 type II collagen; AOM; Cartilage collagen; Chondrocalcin; CO2A1_HUMAN; COL11A3; Col2a1; Collagen 2; Collagen II alpha 1 polypeptide; SEDC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7367