Learn More
Biomatik Corporation Human Collagen Type Vi (Colvi) Elisa Kit
Supplier:  Biomatik Corporation EKC33227
Encompass_Preferred
Human Collagen Type Vi (Colvi) Elisa Kit for detecting Collagen, Type Vi, Alpha 1 in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12109; COL6A1. Target Synonyms: Alpha 1 (VI) chain (61 AA); CO6A1_HUMAN; COL6A1; COL6A2; COL6A3; collagen 6; Collagen alpha 2 (VI) chain; Collagen alpha 3 (VI) chain; Collagen alpha-1 (VI) chain; collagen six; Collagen type VI alpha 1; Collagen type VI alpha 2; Collagen type VI alpha 3; Collagen VI alpha 1 polypeptide; Collagen VI alpha 2 polypeptide; Collagen VI alpha 3 polypeptide; CollagenVI; Human mRNA for collagen VI alpha 2 C terminal globular domain; OPLL; PP3610. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7373