Learn More
Biomatik Corporation Cattle Resistin (RETN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07082
Encompass_Preferred
The Cattle Resistin (RETN) ELISA Kit allows for quantitative determination of Resistin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 78-5000 pg/mL. Sensitivity: 31 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q762I5. Cross Activity: No significant cross-reactivity or interference between Resistin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3692