Learn More
Biomatik Corporation Human Fibrinogen (Fb) Elisa Kit
Supplier:  Biomatik Corporation EKC33684
Encompass_Preferred
Human Fibrinogen (Fb) Elisa Kit for detecting Fibrinogen(Fb) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ug/ml. Sensitivity: 0.078ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7416