Learn More
Biomatik Corporation Human Resistin Like Beta (RETNLb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07086
Encompass_Preferred
Human Resistin Like Beta (RETNLb) ELISA Kit for detecting RETNLb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 21.3ppg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQ08. Gene ID: 84666. Target Synonyms: XCP2; FIZZ2; HXCP2; CCRG; RETNL2; RELMb; Foundin In Flammatory Zone 2; Colon and small intestine-specific cysteine-rich protein; Colon carcinoma-related gene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3695