Learn More
Biomatik Corporation Human Insulin-Like Growth Factors 1, Igf-1 Elisa Kit
Supplier:  Biomatik Corporation EKC34153
Encompass_Preferred
Human Insulin-Like Growth Factors 1, Igf-1 Elisa Kit for detecting Insulin-Like Growth Factors 1, Igf-1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05019; IGF1. Target Synonyms: IBP1; IGF I; IGF IA; IGF IB; IGF-I; Igf1; IGF1_HUMAN; IGF1A; IGFI; IGFIA; Insulin like growth factor 1 (somatomedin C); Insulin like growth factor 1; Insulin like growth factor IA; Insulin like growth factor IB; Insulin-like growth factor I; Mechano growth factor; MGF; OTTHUMP00000195080; OTTHUMP00000195081; OTTHUMP00000195082; OTTHUMP00000195083; OTTHUMP00000195084; Somatomedia C; Somatomedin C; Somatomedin-C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7445