Learn More
Biomatik Corporation Human Retinoic Acid Receptor Alpha (RARa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07093
Encompass_Preferred
Human Retinoic Acid Receptor Alpha (RARa) ELISA Kit for detecting RARa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10276. Gene ID: 5914. Target Synonyms: RAR-A; NR1B1; RAR; RAR-Alpha; Nuclear Receptor Subfamily 1 Group B Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3699