Learn More
Biomatik Corporation Human Neural -Cadherin, N-Cad Elisa Kit
Supplier:  Biomatik Corporation EKC34765
Encompass_Preferred
Human Neural -Cadherin, N-Cad Elisa Kit for detecting Cadherin 2, Type 1, N-Cadherin (Neuronal) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19022; CDH2. Target Synonyms: CDH2; CDHN; NCAD; Cadherin-2; CDw325; Neural cadherin; N-cadherin; CD antigen CD325. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7489