Learn More
Biomatik Corporation Mouse Retinol Binding Protein 4, Plasma (RBP4) ELISA Kit, 96 tests
Catalog No. 501503703
Encompass_Preferred
Mouse Retinol Binding Protein 4 (RBP4) ELISA Kit for detecting RBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.142ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00724. Gene ID: 19662. Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3703 Supplier Biomatik Corporation Supplier No. EKU07105
Mouse Retinol Binding Protein 4 (RBP4) ELISA Kit for detecting RBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.142ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00724. Gene ID: 19662. Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.