Learn More
Biomatik Corporation Human protein(peptidylprolyl cis/trans isomerase)NIMA-interacting 1,PIN1 ELISA Kit
Supplier:  Biomatik Corporation EKC35263
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q13526. Target Synonyms: DOD, UBL5, peptidyl-prolyl cis/trans isomerase, NIMA-interacting|prolyl isomerase|protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1|protein (peptidylprolyl cis/trans isomerase) NIMA-i. Restriction: For Research Use Only.
Catalog No. 50-228-7530