Learn More
Biomatik Corporation Human Retinoschisin (RS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07107
Encompass_Preferred
Human Retinoschisin (RS) ELISA Kit for detecting RS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15537. Gene ID: 6247. Target Synonyms: RS1; XLRS1; Retinoschisis X-Linked,Juvenile 1; X-linked juvenile retinoschisis protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3704