Learn More
Biomatik Corporation Human Retinol Binding Protein 4, Rbp-4 Elisa Kit
Supplier:  Biomatik Corporation EKC35385
Encompass_Preferred
Human Retinol Binding Protein 4, Rbp-4 Elisa Kit for detecting Retinol Binding Protein 4, Plasma in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02753; RBP4. Target Synonyms: OTTHUMP00000020114; OTTHUMP00000020115; OTTHUMP00000020116; Plasma retinol binding protein 4; Plasma retinol-binding protein; Plasma retinol-binding protein (1-176); PRBP; PRO2222; RBP; RBP4; RDCCAS; RET4_HUMAN; Retinol binding protein 4; Retinol binding protein 4 interstitial; Retinol binding protein 4 plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7537