Learn More
Biomatik Corporation Human Rho GDP Dissociation Inhibitor Beta (ARHGDIb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07113
Encompass_Preferred
Human Rho GDP Dissociation Inhibitor Beta (ARHGDIb) ELISA Kit for detecting ARHGDIb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.241ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52566. Gene ID: 397. Target Synonyms: D4; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; RhoGDI2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3706