Learn More
Biomatik Corporation Cattle Ribonuclease A (RNASE1) ELISA Kit, 96 tests
Catalog No. 501503707
Encompass_Preferred
The Cattle Ribonuclease A (RNASE1) ELISA Kit allows for quantitative determination of Ribonuclease A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P61823. Cross Activity: No significant cross-reactivity or interference between Ribonuclease A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3707 Supplier Biomatik Corporation Supplier No. EKU07118
The Cattle Ribonuclease A (RNASE1) ELISA Kit allows for quantitative determination of Ribonuclease A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P61823. Cross Activity: No significant cross-reactivity or interference between Ribonuclease A and analogues was observed. Restriction: For Research Use Only.