Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 2, Bmp-2 Elisa Kit
Supplier:  Biomatik Corporation EKC36417
Encompass_Preferred
Mouse Bone Morphogenetic Protein 2, Bmp-2 Elisa Kit for detecting Bone Morphogenetic Protein 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21274; Bmp2. Target Synonyms: Bmp2; Bmp-2; Bone morphogenetic protein 2; BMP-2; Bone morphogenetic protein 2A; BMP-2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7576