Learn More
Biomatik Corporation Human Ribonuclease A2 (RNASE2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07120
Encompass_Preferred
Human Ribonuclease A2 (RNASE2) ELISA Kit for detecting RNASE2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10153. Gene ID: 6036. Target Synonyms: Rnase-A2; Rnase2; EDN; Ribonuclease US; Ribonuclease,RNase A Family,2; Liver,Eosinophil-Derived Neurotoxin; MNon-secretory ribonuclease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3709