Learn More
Biomatik Corporation Mouse interferon-inducible protein 10,IP-10 ELISA Kit
Supplier:  Biomatik Corporation EKC37145
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/ml - 20 ng/ml. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P17515. Target Synonyms: C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1, gamma IP10|interferon-inducible cytokine IP-10|protein 10 from interferon (gamma)-induced cell line|small inducible cytokine B10|small inducibl. Restriction: For Research Use Only.
Catalog No. 50-228-7603