Learn More
Biomatik Corporation Human Ribonuclease A6 (RNASE6) ELISA Kit, 96 tests
Catalog No. 501503711
Encompass_Preferred
Human Ribonuclease A6 (RNASE6) ELISA Kit for detecting RNASE6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q93091. Gene ID: 6039. Target Synonyms: RNS6; Rnase-A6; RNase K6; Ribonuclease,RNase A Family,K6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3711 Supplier Biomatik Corporation Supplier No. EKU07122
Human Ribonuclease A6 (RNASE6) ELISA Kit for detecting RNASE6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q93091. Gene ID: 6039. Target Synonyms: RNS6; Rnase-A6; RNase K6; Ribonuclease,RNase A Family,K6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.