Learn More
Biomatik Corporation Pig Angiopoietin 1,ANG-1 ELISA Kit
Supplier:  Biomatik Corporation EKC38091
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Sus scrofa (Pig). Detection Range: 6 ng/mL-200 ng/mL. Sensitivity: 6 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BDY8. Target Synonyms: AGP1, AGPT, ANG1. Restriction: For Research Use Only.
Catalog No. 50-228-7629