Learn More
Biomatik Corporation Rabbit Interleukin 4,IL-4 ELISA Kit
Supplier:  Biomatik Corporation EKC38472
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.625 pg/mL-40 pg/mL. Sensitivity: 0.156 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9MZR8. Target Synonyms: BCGF-1, BCGF1, BSF1, IL-4, MGC79402, B cell growth factor 1|B_cell stimulatory factor 1|lymphocyte stimulatory factor 1. Restriction: For Research Use Only.
Catalog No. 50-228-7644