Learn More
Biomatik Corporation Human Ribonuclease L (RNASEL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07127
Encompass_Preferred
Human Ribonuclease L (RNASEL) ELISA Kit for detecting RNASEL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05823. Gene ID: 6041. Target Synonyms: Rnase-L; RNS4; PRCA1; 2',5'-Oligoisoadenylate Synthetase-Dependent; Prostate Cancer 1; 2-5A-dependent ribonuclease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3713