Learn More
Biomatik Corporation Rat Endothelial Nitric Oxide Synthase, Enos/Nos3 Elisa Kit
Supplier:  Biomatik Corporation EKC38987
Encompass_Preferred
Rat Endothelial Nitric Oxide Synthase, Enos/Nos3 Elisa Kit for detecting Nitric Oxide Synthase 3 (Endothelial Cell) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000uIU/ml. Sensitivity: 3.9uIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q62600; Nos3. Target Synonyms: Nos3; Nitric oxide synthase; endothelial; EC 1.14.13.39; Constitutive NOS; cNOS; EC-NOS; Endothelial NOS; eNOS; NOS type III; NOSIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7658